missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobiphenyl, 97+%

CAS: 1591-31-7 | C12H9I | 280.11 g/mol

$255.42 - $255.42

Chemical Identifiers

CAS 1591-31-7
Molecular Formula C12H9I
Molecular Weight (g/mol) 280.11
MDL Number MFCD00019028
InChI Key NXYICUMSYKIABQ-UHFFFAOYSA-N
Synonym 4-iodobiphenyl, 4-iodo-1,1'-biphenyl, 1,1'-biphenyl, 4-iodo, p-iodobiphenyl, 4-iododiphenyl, p-phenyliodobenzene, 4-biphenyl iodide, biphenyl, 4-iodo, 1-iodo-4-phenyl-benzene, 4-iodo-1-phenylbenzene
PubChem CID 15322
IUPAC Name 1-iodo-4-phenylbenzene
SMILES IC1=CC=C(C=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC392390050
SDS View Documents
Thermo Scientific Chemicals
392390050
5 g Glass bottle
Each for $255.42
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1591-31-7
280.11
NXYICUMSYKIABQ-UHFFFAOYSA-N
15322
IC1=CC=C(C=C1)C1=CC=CC=C1
C12H9I
MFCD00019028
4-iodobiphenyl, 4-iodo-1,1'-biphenyl, 1,1'-biphenyl, 4-iodo, p-iodobiphenyl, 4-iododiphenyl, p-phenyliodobenzene, 4-biphenyl iodide, biphenyl, 4-iodo, 1-iodo-4-phenyl-benzene, 4-iodo-1-phenylbenzene
1-iodo-4-phenylbenzene

Specifications

1591-31-7
100.0
White
97% min. (HPLC)
C12H9I
MFCD00019028
4-iodobiphenyl, 4-iodo-1,1'-biphenyl, 1,1'-biphenyl, 4-iodo, p-iodobiphenyl, 4-iododiphenyl, p-phenyliodobenzene, 4-biphenyl iodide, biphenyl, 4-iodo, 1-iodo-4-phenyl-benzene, 4-iodo-1-phenylbenzene
IC1=CC=C(C=C1)C1=CC=CC=C1
280.11
280.1
Crystalline Powder
97.0
113°C
Authentic
Glass bottle
C6H5C6H4I
5 g
NXYICUMSYKIABQ-UHFFFAOYSA-N
1-iodo-4-phenylbenzene
15322
97+%
4-Iodobiphenyl
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-469-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only