missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobiphenyl, 97+%

Catalog No. AC392390010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392390010 1 g Glass bottle
AC392390050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC392390010 Supplier Thermo Scientific Chemicals Supplier No. 392390010

CAS: 1591-31-7 | C12H9I | 280.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1591-31-7
Molecular Formula C12H9I
Molecular Weight (g/mol) 280.11
MDL Number MFCD00019028
InChI Key NXYICUMSYKIABQ-UHFFFAOYSA-N
Synonym 4-iodobiphenyl, 4-iodo-1,1'-biphenyl, 1,1'-biphenyl, 4-iodo, p-iodobiphenyl, 4-iododiphenyl, p-phenyliodobenzene, 4-biphenyl iodide, biphenyl, 4-iodo, 1-iodo-4-phenyl-benzene, 4-iodo-1-phenylbenzene
PubChem CID 15322
IUPAC Name 1-iodo-4-phenylbenzene
SMILES IC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 113°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5C6H4I
Quantity 1 g
Formula Weight 280.1
Percent Purity 97+%
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodobiphenyl
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-469-1

RUO – Research Use Only