Learn More
4-Iodobiphenyl, 97%
CAS: 1591-31-7 | C12H9I | 280.11 g/mol
Supplier: Thermo Scientific Chemicals H2745306
Description
4-Iodobiphenyl is used as a starting material for the preparation of p-terphenyl derivatives. It is also used in Suzuki reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Iodobiphenyl is used as a starting material for the preparation of p-terphenyl derivatives. It is also used in Suzuki reaction.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases.
Specifications
| 4-Iodobiphenyl | |
| 110°C to 114°C | |
| MFCD00019028 | |
| UN3077 | |
| 4-iodobiphenyl, 4-iodo-1,1'-biphenyl, 1,1'-biphenyl, 4-iodo, p-iodobiphenyl, 4-iododiphenyl, p-phenyliodobenzene, 4-biphenyl iodide, biphenyl, 4-iodo, 1-iodo-4-phenyl-benzene, 4-iodo-1-phenylbenzene | |
| NXYICUMSYKIABQ-UHFFFAOYSA-N | |
| 1-iodo-4-phenylbenzene | |
| 15322 | |
| 97% |
| 1591-31-7 | |
| C12H9I | |
| 5 g | |
| Light sensitive | |
| Slightly soluble in water. | |
| IC1=CC=C(C=C1)C1=CC=CC=C1 | |
| 280.11 | |
| 280.11 |
Chemical Identifiers
| 1591-31-7 | |
| 280.11 | |
| NXYICUMSYKIABQ-UHFFFAOYSA-N | |
| 15322 | |
| IC1=CC=C(C=C1)C1=CC=CC=C1 |
| C12H9I | |
| MFCD00019028 | |
| 4-iodobiphenyl, 4-iodo-1,1'-biphenyl, 1,1'-biphenyl, 4-iodo, p-iodobiphenyl, 4-iododiphenyl, p-phenyliodobenzene, 4-biphenyl iodide, biphenyl, 4-iodo, 1-iodo-4-phenyl-benzene, 4-iodo-1-phenylbenzene | |
| 1-iodo-4-phenylbenzene |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 216-469-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only