missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzylamine hydrochloride, 97%

Catalog No. AAL1996706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1996706 5 g
AAL1996703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1996706 Supplier Thermo Scientific Chemicals Supplier No. L1996706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59528-27-7 | C7H9ClIN | 269.51 g/mol

4-Iodobenzylamine hydrochloride is used to prepare cyclic amide by reacting with methyl 4-bromomethyl-3-methoxycarbonylcinnamate in the presence of triethylamine. It is also used in pharmaceuticals, agrochemicals and dye industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzylamine hydrochloride is used to prepare cyclic amide by reacting with methyl 4-bromomethyl-3-methoxycarbonylcinnamate in the presence of triethylamine. It is also used in pharmaceuticals, agrochemicals and dye industries.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 59528-27-7
Molecular Formula C7H9ClIN
Molecular Weight (g/mol) 269.51
MDL Number MFCD04039324
InChI Key GBJMURRFWZREHE-UHFFFAOYSA-N
Synonym 4-iodobenzylamine hydrochloride, 4-iodophenyl methanamine hydrochloride, 4-iodobenzylamine hcl, benzenemethanamine, 4-iodo-, hydrochloride, 4-iodophenyl methylamine, chloride, 1-4-iodophenyl methanamine hydrochloride, 4-iodobenzylamine, hydrochloride salt, 4-iodobenzylaminehydrochloride, pubchem3304, acmc-209tzk
PubChem CID 16217505
IUPAC Name (4-iodophenyl)methanamine;hydrochloride
SMILES C1=CC(=CC=C1CN)I.Cl

Specifications

Melting Point 275°C to 280°C (decomposition)
Quantity 5 g
Beilstein 3691299
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 269.51
Percent Purity 97%
Chemical Name or Material 4-Iodobenzylamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only