missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodobenzoyl Chloride 98.0+%, TCI America™
$98.79 - $287.06
Chemical Identifiers
| CAS | 1711-02-0 |
|---|---|
| Molecular Formula | C7H4ClIO |
| Molecular Weight (g/mol) | 266.462 |
| MDL Number | MFCD00001058 |
| InChI Key | NJAKCIUOTIPYED-UHFFFAOYSA-N |
| Synonym | p-iodobenzoyl chloride, benzoyl chloride, 4-iodo, benzoyl chloride, p-iodo, 4-iodobenzoic acid chloride, 4-iodobenzoylchloride, 4-iodo-benzoyl chloride, 4iodobenzoyl chloride, 4-iodo-benzoylchlorid, rho-iodobenzoyl chloride, 4-iodo benzoyl chloride |
| PubChem CID | 74373 |
| IUPAC Name | 4-iodobenzoyl chloride |
| SMILES | C1=CC(=CC=C1C(=O)Cl)I |
Chemical Identifiers
| 1711-02-0 | |
| 266.462 | |
| NJAKCIUOTIPYED-UHFFFAOYSA-N | |
| 74373 | |
| C1=CC(=CC=C1C(=O)Cl)I |
| C7H4ClIO | |
| MFCD00001058 | |
| p-iodobenzoyl chloride, benzoyl chloride, 4-iodo, benzoyl chloride, p-iodo, 4-iodobenzoic acid chloride, 4-iodobenzoylchloride, 4-iodo-benzoyl chloride, 4iodobenzoyl chloride, 4-iodo-benzoylchlorid, rho-iodobenzoyl chloride, 4-iodo benzoyl chloride | |
| 4-iodobenzoyl chloride |
Specifications
| 1711-02-0 | |
| White-Yellow | |
| C7H4ClIO | |
| 5 g | |
| p-iodobenzoyl chloride, benzoyl chloride, 4-iodo, benzoyl chloride, p-iodo, 4-iodobenzoic acid chloride, 4-iodobenzoylchloride, 4-iodo-benzoyl chloride, 4iodobenzoyl chloride, 4-iodo-benzoylchlorid, rho-iodobenzoyl chloride, 4-iodo benzoyl chloride | |
| C1=CC(=CC=C1C(=O)Cl)I | |
| 266.462 | |
| 266.46 | |
| Crystalline Powder |
| 65°C | |
| 100°C | |
| MFCD00001058 | |
| 3261 | |
| NJAKCIUOTIPYED-UHFFFAOYSA-N | |
| 4-iodobenzoyl chloride | |
| 74373 | |
| ≥98.0% (GC,T) | |
| 4-Iodobenzoyl Chloride |
Safety and Handling
TSCA : Yes