missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzoyl Chloride 98.0+%, TCI America™

Catalog No. I05085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05085G 5 g
I050825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05085G Supplier TCI America Supplier No. I05085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1711-02-0
Molecular Formula C7H4ClIO
Molecular Weight (g/mol) 266.462
MDL Number MFCD00001058
InChI Key NJAKCIUOTIPYED-UHFFFAOYSA-N
Synonym p-iodobenzoyl chloride, benzoyl chloride, 4-iodo, benzoyl chloride, p-iodo, 4-iodobenzoic acid chloride, 4-iodobenzoylchloride, 4-iodo-benzoyl chloride, 4iodobenzoyl chloride, 4-iodo-benzoylchlorid, rho-iodobenzoyl chloride, 4-iodo benzoyl chloride
PubChem CID 74373
IUPAC Name 4-iodobenzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)I

Specifications

Melting Point 65°C
Color White-Yellow
Boiling Point 100°C
Quantity 5 g
UN Number 3261
Formula Weight 266.46
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodobenzoyl Chloride