missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzoyl chloride, 98%

Catalog No. AC412390250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412390250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC412390250 Supplier Thermo Scientific Chemicals Supplier No. 412390250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1711-02-0 | C7H4ClIO | 266.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1711-02-0
Molecular Formula C7H4ClIO
Molecular Weight (g/mol) 266.46
MDL Number MFCD00001058
InChI Key NJAKCIUOTIPYED-UHFFFAOYSA-N
Synonym p-iodobenzoyl chloride, benzoyl chloride, 4-iodo, benzoyl chloride, p-iodo, 4-iodobenzoic acid chloride, 4-iodobenzoylchloride, 4-iodo-benzoyl chloride, 4iodobenzoyl chloride, 4-iodo-benzoylchlorid, rho-iodobenzoyl chloride, 4-iodo benzoyl chloride
PubChem CID 74373
IUPAC Name 4-iodobenzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)I

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 62°C to 66°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4COCl
Quantity 25 g
Beilstein 09, 367
Formula Weight 266.46
Percent Purity 98%
Physical Form Crystalline Mass or Crystalline Powder and Chunks
Chemical Name or Material 4-Iodobenzoyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-974-7
TSCA TSCA

RUO – Research Use Only