missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzotrifluoride, 98%

CAS: 455-13-0 | C7H4F3I | 272.009 g/mol

$48.19 - $529.59

Chemical Identifiers

CAS 455-13-0
Molecular Formula C7H4F3I
Molecular Weight (g/mol) 272.009
MDL Number MFCD00039398
InChI Key SKGRFPGOGCHDPC-UHFFFAOYSA-N
Synonym 4-iodobenzotrifluoride, 1-iodo-4-trifluoromethyl benzene, p-iodobenzotrifluoride, benzene, 1-iodo-4-trifluoromethyl, 4-trifluoromethyl iodobenzene, 4-iodo-alpha,alpha,alpha-trifluorotoluene, alpha,alpha,alpha-trifluoro-4-iodotoluene, 4-iodobenztrifluoride, 4-iodobenzo trifluoride, 4-iodo-1-trifluoromethylbenzene
PubChem CID 67993
IUPAC Name 1-iodo-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)I
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1218903
View Documents
Thermo Scientific Chemicals
A1218903
1 g
Each for $48.19
Only null left
 
AAA1218906
View Documents
Thermo Scientific Chemicals
A1218906
5 g
Each for $158.36
Only null left
 
AAA1218914
View Documents
Thermo Scientific Chemicals
A1218914
25 g
Each for $529.59
Only null left
 
Description

Description

4-Iodobenzotrifluoride may be employed as substrate with electron-deficient aromatic ring, during Mizoroki-Heck reaction with acrylic acid, to afford 4-trifluoromethylcinnnamic acid. It undergoes aminocarbonylation in DMF using phosphoryl chloride to give N,N-dimethyl-(4-trifluoromethyl)benzamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzotrifluoride may be employed as substrate with electron-deficient aromatic ring, during Mizoroki-Heck reaction with acrylic acid, to afford 4-trifluoromethylcinnnamic acid. It undergoes aminocarbonylation in DMF using phosphoryl chloride to give N,N-dimethyl-(4-trifluoromethyl)benzamide.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents. Store in dark.
Specifications

Chemical Identifiers

455-13-0
272.009
SKGRFPGOGCHDPC-UHFFFAOYSA-N
67993
C1=CC(=CC=C1C(F)(F)F)I
C7H4F3I
MFCD00039398
4-iodobenzotrifluoride, 1-iodo-4-trifluoromethyl benzene, p-iodobenzotrifluoride, benzene, 1-iodo-4-trifluoromethyl, 4-trifluoromethyl iodobenzene, 4-iodo-alpha,alpha,alpha-trifluorotoluene, alpha,alpha,alpha-trifluoro-4-iodotoluene, 4-iodobenztrifluoride, 4-iodobenzo trifluoride, 4-iodo-1-trifluoromethylbenzene
1-iodo-4-(trifluoromethyl)benzene

Specifications

455-13-0
1.851
68°C (154°F)
1.518
1 g
Light sensitive
Insoluble in water.
C1=CC(=CC=C1C(F)(F)F)I
272.009
272.01
4-Iodobenzotrifluoride
-8°C
68°C to 70°C (15 mmHg)
C7H4F3I
MFCD00039398
1944013
4-iodobenzotrifluoride, 1-iodo-4-trifluoromethyl benzene, p-iodobenzotrifluoride, benzene, 1-iodo-4-trifluoromethyl, 4-trifluoromethyl iodobenzene, 4-iodo-alpha,alpha,alpha-trifluorotoluene, alpha,alpha,alpha-trifluoro-4-iodotoluene, 4-iodobenztrifluoride, 4-iodobenzo trifluoride, 4-iodo-1-trifluoromethylbenzene
SKGRFPGOGCHDPC-UHFFFAOYSA-N
1-iodo-4-(trifluoromethyl)benzene
67993
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 207-234-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only