missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7131183
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
I00545G 5 g
I005425G 25 g
2 options

Catalog No. I00545G

Supplier: TCI America I00545G

Only null left

Chemical Identifiers

CAS 619-58-9
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.02
MDL Number MFCD00002533
InChI Key GHICCUXQJBDNRN-UHFFFAOYSA-N
Synonym p-iodobenzoic acid, benzoic acid, 4-iodo, benzoic acid, p-iodo, 4-iodo-benzoic acid, p-iodobenzenecarboxylic acid, 4-jodbenzoesaeure, unii-ipo4lyq1en, 4-jodbenzoesaeure german, 4-iodo benzoic acid, ipo4lyq1en
PubChem CID 12085
IUPAC Name 4-iodobenzoic acid
SMILES OC(=O)C1=CC=C(I)C=C1

Specifications

Quantity 5 g
Formula Weight 248.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodobenzoic Acid
RTECSNumber DH2976000
TSCA Yes