missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzoic acid, 97%

Catalog No. p-7080931
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1073009 10 g
AAA1073018 50 g
AAA1073030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1073009 Supplier Thermo Scientific Chemicals Supplier No. A1073009
Only null left

CAS: 619-58-9 | C7H5IO2 | 248.02 g/mol

4-Iodobenzoic acid was used in the synthesis of [hydroxy(4-carboxyphenyl)iodonium]ion in situ that helps in the cleavage of a variety of alkenes. It is is used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzoic acid was used in the synthesis of [hydroxy(4-carboxyphenyl)iodonium]ion in situ that helps in the cleavage of a variety of alkenes. It is is used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

Solubility
Partly miscible with water. Soluble in alcohol & ether.

Notes
It is sensitive to light. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 619-58-9
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.02
MDL Number MFCD00002533
InChI Key GHICCUXQJBDNRN-UHFFFAOYSA-N
Synonym p-iodobenzoic acid, benzoic acid, 4-iodo, benzoic acid, p-iodo, 4-iodo-benzoic acid, p-iodobenzenecarboxylic acid, 4-jodbenzoesaeure, unii-ipo4lyq1en, 4-jodbenzoesaeure german, 4-iodo benzoic acid, ipo4lyq1en
PubChem CID 12085
IUPAC Name 4-iodobenzoic acid
SMILES OC(=O)C1=CC=C(I)C=C1

Specifications

Melting Point ∼265°C (decomposition)
Density 2.18
Quantity 10 g
Beilstein 1860232
Sensitivity Light sensitive
Solubility Information Partly miscible with water. Soluble in alcohol & ether.
Formula Weight 248.02
Percent Purity 97%
Chemical Name or Material 4-Iodobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-603-2
RTECSNumber DH2976000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only