missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzeneboronic acid, 97%

Catalog No. AAH5111903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5111903 1 g
AAH5111906 5 g
AAH5111914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5111903 Supplier Thermo Scientific Chemicals Supplier No. H5111903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5122-99-6 | C6H6BIO2 | 247.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5122-99-6
Molecular Formula C6H6BIO2
Molecular Weight (g/mol) 247.83
MDL Number MFCD01319014
InChI Key PELJYVULHLKXFF-UHFFFAOYSA-N
Synonym 4-iodophenyl boronic acid, 4-iodobenzeneboronic acid, p-iodophenylboronic acid, 4-iodophenylboronicacid, p-iodobenzeneboronic acid, p-iodophenyl boronic acid, boronic acid, 4-iodophenyl, 4-boronoiodobenzene, pubchem6160, pubchem11574
PubChem CID 151254
IUPAC Name (4-iodophenyl)boronic acid
SMILES OB(O)C1=CC=C(I)C=C1

Specifications

Quantity 1 g
Sensitivity Light sensitive
Formula Weight 247.83
Percent Purity 97%
Chemical Name or Material 4-Iodobenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only