missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzaldehyde, 98+%

Catalog No. p-7047045
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL17448MD 250 mg
AAL1744803 1 g
AAL1744806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL17448MD Supplier Thermo Scientific Chemicals Supplier No. L17448MD

CAS: 15164-44-0 | C7H5IO | 232.02 g/mol

4-Iodobenzaldehyde is used in synthesis of 4-[2-(trimethylsilyl)ethynyl]benzaldehyde, 5,15-dimesityl-10-(3-[2-(trimethylsilyl)ethynyi]phenyl}-20-(4-iodophenyl)porphyrin, and 5,15-dimesityl-10-[3,5-bis{2-[4-(N,N?-difluoroboryl-1,9-dimethyidipyrrin-5-yl)-phenyl]ethynyl}phenyl]-20-(4-iodophenyl)porphyrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzaldehyde is used in synthesis of 4-[2-(trimethylsilyl)ethynyl]benzaldehyde, 5,15-dimesityl-10-(3-[2-(trimethylsilyl)ethynyi]phenyl}-20-(4-iodophenyl)porphyrin, and 5,15-dimesityl-10-[3,5-bis{2-[4-(N,N′-difluoroboryl-1,9-dimethyidipyrrin-5-yl)-phenyl]ethynyl}phenyl]-20-(4-iodophenyl)porphyrin.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air and light. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 15164-44-0
Molecular Formula C7H5IO
Molecular Weight (g/mol) 232.02
MDL Number MFCD00039576
InChI Key NIEBHDXUIJSHSL-UHFFFAOYSA-N
Synonym p-iodobenzaldehyde, benzaldehyde, 4-iodo, 4-iodo-benzaldehyde, iodobenzenecarbaldehyde, 4-iodo benzaldehyde, pubchem5349, acmc-1bpp9, intermediates-zcf02160, 4-iodobenzaldehyde, ksc181a1r
PubChem CID 96657
IUPAC Name 4-iodobenzaldehyde
SMILES IC1=CC=C(C=O)C=C1

Specifications

Melting Point 76°C to 78°C
Quantity 250 mg
Beilstein 636101
Sensitivity Air and light sensitive
Solubility Information Insoluble in water.
Formula Weight 232.02
Percent Purity ≥98%
Chemical Name or Material 4-Iodobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only