missing translation for 'onlineSavingsMsg'
Learn More

4-Iodoanisole 99.0+%, TCI America™

Catalog No. I037825G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I037825G 25 g
I0378100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I037825G Supplier TCI America Supplier No. I037825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 696-62-8
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD00001056
InChI Key SYSZENVIJHPFNL-UHFFFAOYSA-N
Synonym 4-iodoanisole, p-iodoanisole, benzene, 1-iodo-4-methoxy, isoform, 4-methoxyiodobenzene, p-methoxyiodobenzene, anisole, p-iodo, 4-iodomethoxybenzene, p-iodophenyl methyl ether, 1-iodo-4-methoxy-benzene
PubChem CID 69676
IUPAC Name 1-iodo-4-methoxybenzene
SMILES COC1=CC=C(C=C1)I

Specifications

Melting Point 51°C
Boiling Point 240°C
Quantity 25 g
Formula Weight 234.04
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodoanisole
EINECSNumber (3)-3916
TSCA Yes