missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodoanisole, 98%
CAS: 696-62-8 | C7H7IO | 234.04 g/mol
$73.37 - $73.37
Chemical Identifiers
| CAS | 696-62-8 |
|---|---|
| Molecular Formula | C7H7IO |
| Molecular Weight (g/mol) | 234.04 |
| InChI Key | SYSZENVIJHPFNL-UHFFFAOYSA-N |
| Synonym | 4-iodoanisole, p-iodoanisole, benzene, 1-iodo-4-methoxy, isoform, 4-methoxyiodobenzene, p-methoxyiodobenzene, anisole, p-iodo, 4-iodomethoxybenzene, p-iodophenyl methyl ether, 1-iodo-4-methoxy-benzene |
| PubChem CID | 69676 |
| IUPAC Name | 1-iodo-4-methoxybenzene |
| SMILES | COC1=CC=C(C=C1)I |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC122320250
|
Thermo Scientific Chemicals
122320250 |
25 g | Glass bottle |
Each for $73.37
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 696-62-8 | |
| 234.04 | |
| 4-iodoanisole, p-iodoanisole, benzene, 1-iodo-4-methoxy, isoform, 4-methoxyiodobenzene, p-methoxyiodobenzene, anisole, p-iodo, 4-iodomethoxybenzene, p-iodophenyl methyl ether, 1-iodo-4-methoxy-benzene | |
| 1-iodo-4-methoxybenzene |
| C7H7IO | |
| SYSZENVIJHPFNL-UHFFFAOYSA-N | |
| 69676 | |
| COC1=CC=C(C=C1)I |
Specifications
| 696-62-8 | |
| Brown to White | |
| >110°C | |
| 97.5% min. (GC) | |
| C7H7IO | |
| 25 g | |
| 4-iodoanisole, p-iodoanisole, benzene, 1-iodo-4-methoxy, isoform, 4-methoxyiodobenzene, p-methoxyiodobenzene, anisole, p-iodo, 4-iodomethoxybenzene, p-iodophenyl methyl ether, 1-iodo-4-methoxy-benzene | |
| COC1=CC=C(C=C1)I | |
| 234.04 | |
| 234.04 | |
| Crystalline Powder and/or Chunks |
| 46.0°C to 51.0°C | |
| 237.0°C (726.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| IC6H4OCH3 | |
| 06,208 | |
| SYSZENVIJHPFNL-UHFFFAOYSA-N | |
| 1-iodo-4-methoxybenzene | |
| 69676 | |
| 98% | |
| 4-Iodoanisole |
Safety and Handling
EINECSNumber : 211-798-7
Recommended Storage : Product may darken on storage
RUO – Research Use Only