missing translation for 'onlineSavingsMsg'
Learn More

4-Iodoaniline 99.0+%, TCI America™

Catalog No. I004825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I004825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I004825G Supplier TCI America Supplier No. I004825G

missing translation for 'validMainframeMsg'

Specifications

CAS 540-37-4
Melting Point 64°C
Molecular Formula C6H6IN
Quantity 25 g
MDL Number MFCD00007848
Synonym p-iodoaniline, benzenamine, 4-iodo, 4-iodobenzenamine, aniline, p-iodo, p-aminophenyl iodide, aniline, 4-iodo, 4-iodo-aniline, 4-iodophenylamine, 4-iodo-benzenamine, 4-iodo-phenylamine
InChI Key VLVCDUSVTXIWGW-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1N)I
IUPAC Name 4-iodoaniline
Molecular Weight (g/mol) 219.025
PubChem CID 10893
Formula Weight 219.03
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodoaniline
Show More Show Less
RTECSNumber BY3850000
TSCA Yes