missing translation for 'onlineSavingsMsg'
Learn More

4'-Iodoacetanilide 95.0+%, TCI America™

Catalog No. I08461G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08461G 1 g
I08465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08461G Supplier TCI America Supplier No. I08461G

Chemical Identifiers

CAS 622-50-4
Molecular Formula C8H8INO
Molecular Weight (g/mol) 261.062
MDL Number MFCD00016352
InChI Key SIULLDWIXYYVCU-UHFFFAOYSA-N
Synonym n-4-iodophenyl acetamide, 4-iodoacetanilide, 4'-iodoacetanilide, p-iodoacetanilide, acetamide, n-4-iodophenyl, acetanilide, 4'-iodo, 4'-iodo-acetanilide, pubchem3293, acmc-20akb8, maybridge1_006480
PubChem CID 12147
IUPAC Name N-(4-iodophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)I

Specifications

Melting Point 183°C
Quantity 1 g
Formula Weight 261.06
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Iodoacetanilide
RTECSNumber AE4285000
TSCA Yes