accessibility menu, dialog, popup

UserName

CH3ONHI

CAS RN 622-50-4

IUPAC Name: N-(4-iodophenyl)acetamide
Synonyms: 4-iodoacetanilide
Molecular Weight (g/mol): 261.06
Molecular Formula: C8H8INO
InChi Key: SIULLDWIXYYVCU-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(I)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)