missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-m-xylene, 98%

Catalog No. AAA1687818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1687818 50 g
AAA1687809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1687818 Supplier Thermo Scientific Chemicals Supplier No. A1687818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4214-28-2 | C8H9I | 232.06 g/mol

4-Iodo-m-xylene is a reactant used in the preparation of paralog-selective Hsp90 inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodo-m-xylene is a reactant used in the preparation of paralog-selective Hsp90 inhibitors.

Solubility
Not miscible or difficult to mix in water.

Notes
Light Sensitive. Protect from light. Store away from oxidizing agents.

Chemical Identifiers

CAS 4214-28-2
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.06
MDL Number MFCD00013706
InChI Key BUNKQJAMHYKQIM-UHFFFAOYSA-N
Synonym 4-iodo-m-xylene, 2,4-dimethyliodobenzene, 1,3-dimethyl-4-iodobenzene, m-xylene, 4-iodo, benzene, 1-iodo-2,4-dimethyl, 2,4-dimethyl-1-iodobenzene, 2-iodo-1,5-dimethylbenzene, 4-jod-m-xylol, pubchem3869, acmc-209jnr
PubChem CID 77885
IUPAC Name 1-iodo-2,4-dimethylbenzene
SMILES CC1=CC=C(I)C(C)=C1

Specifications

Density 1.628
Quantity 50 g
Refractive Index 1.598
Beilstein 1422602
Boiling Point 231°C to 232°C
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 232.07
Percent Purity 98%
Chemical Name or Material 4-Iodo-m-xylene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-141-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only