missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodo-5-methyl-3-phenylisoxazole, 97%, Thermo Scientific™
$119.67 - $119.67
Chemical Identifiers
| CAS | 31295-66-6 |
|---|---|
| Molecular Formula | C10H8INO |
| Molecular Weight (g/mol) | 285.084 |
| MDL Number | MFCD02681888 |
| InChI Key | DDVPBKUHWSZYIP-UHFFFAOYSA-N |
| Synonym | 4-iodo-5-methyl-3-phenylisoxazole, isoxazole, 4-iodo-5-methyl-3-phenyl, 3-phenyl-4-iodo-5-methylisoxazole |
| PubChem CID | 2776150 |
| IUPAC Name | 4-iodo-5-methyl-3-phenyl-1,2-oxazole |
| SMILES | CC1=C(C(=NO1)C2=CC=CC=C2)I |
Chemical Identifiers
| 31295-66-6 | |
| 285.084 | |
| DDVPBKUHWSZYIP-UHFFFAOYSA-N | |
| 2776150 | |
| CC1=C(C(=NO1)C2=CC=CC=C2)I |
| C10H8INO | |
| MFCD02681888 | |
| 4-iodo-5-methyl-3-phenylisoxazole, isoxazole, 4-iodo-5-methyl-3-phenyl, 3-phenyl-4-iodo-5-methylisoxazole | |
| 4-iodo-5-methyl-3-phenyl-1,2-oxazole |
Specifications
| 31295-66-6 | |
| 100.0 | |
| C10H8INO | |
| 1 g | |
| DDVPBKUHWSZYIP-UHFFFAOYSA-N | |
| 4-iodo-5-methyl-3-phenyl-1,2-oxazole | |
| 2776150 | |
| 97% |
| 96.5 | |
| 97% | |
| MFCD02681888 | |
| 4-iodo-5-methyl-3-phenylisoxazole, isoxazole, 4-iodo-5-methyl-3-phenyl, 3-phenyl-4-iodo-5-methylisoxazole | |
| CC1=C(C(=NO1)C2=CC=CC=C2)I | |
| 285.084 | |
| 285.08 | |
| 4-Iodo-5-methyl-3-phenylisoxazole |