missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-3-nitrobenzoic Acid 97.0+%, TCI America™

Catalog No. I09805G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09805G 5 g
I09801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09805G Supplier TCI America Supplier No. I09805G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35674-27-2
Molecular Formula C7H4INO4
Molecular Weight (g/mol) 293.02
MDL Number MFCD00502399
InChI Key DNMTZLCNLAIKQC-UHFFFAOYSA-N
Synonym 4-iodo-3-nitrobenzoic acid, 4-iodo-3-nitrobenzoicacid, 3-nitro-4-iodobenzoic acid, 4-iodo-3-nitro-benzoic acid, benzoic acid, 4-iodo-3-nitro, pubchem13471, acmc-209z8y, 4-iodo-3-nitro benzoic acid
PubChem CID 11208598
IUPAC Name 4-iodo-3-nitrobenzoic acid
SMILES OC(=O)C1=CC(=C(I)C=C1)[N+]([O-])=O

Specifications

Melting Point 212°C
Color Yellow
Quantity 5 g
Formula Weight 293.02
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodo-3-nitrobenzoic Acid