missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-3-nitroanisole, 98+%

Catalog No. AAL1171406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1171406 5 g
AAL1171414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1171406 Supplier Thermo Scientific Chemicals Supplier No. L1171406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 58755-70-7 | C7H6INO3 | 279.033 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58755-70-7
Molecular Formula C7H6INO3
Molecular Weight (g/mol) 279.033
MDL Number MFCD00024195
InChI Key JWZODIRSJJQOKY-UHFFFAOYSA-N
Synonym 4-iodo-3-nitroanisole, 3-nitro-4-iodoanisole, 2-iodo-5-methoxynitrobenzene, 1-iodo-4-methoxy-2-nitro-benzene, benzene, 1-iodo-4-methoxy-2-nitro, 3-nitro-4-iodoanisol, acmc-1b13g, ksc494s9p, 1-iodo-2-nitro-4-methoxybenzene, attercop-chm at115569
PubChem CID 292659
IUPAC Name 1-iodo-4-methoxy-2-nitrobenzene
SMILES COC1=CC(=C(C=C1)I)[N+](=O)[O-]

Specifications

Melting Point 60°C to 63°C
Quantity 5 g
Beilstein 2365592
Sensitivity Light sensitive
Formula Weight 279.04
Percent Purity ≥98%
Chemical Name or Material 4-Iodo-3-nitroanisole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only