missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-3,5-dimethylpyrazole 98.0+%, TCI America™

Catalog No. I07715G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07715G 5 g
I07711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07715G Supplier TCI America Supplier No. I07715G
Only null left

Chemical Identifiers

CAS 2033-45-6
Molecular Formula C5H7IN2
Molecular Weight (g/mol) 222.029
MDL Number MFCD00040247
InChI Key MZZXIXHKDJNBJQ-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-iodopyrazole, 4-iodo-3,5-dimethylpyrazole, 3,5-dimethyl-4-iodo-1h-pyrazole, pyrazole, 3,5-dimethyl-4-iodo, 3,5-dimethyl-4-lodopyrazole, 1h-pyrazole, 4-iodo-3,5-dimethyl, zlchem 469, acmc-209f8u, 5-23-05-00159 beilstein handbook reference, ksc490o8h
PubChem CID 97115
IUPAC Name 4-iodo-3,5-dimethyl-1H-pyrazole
SMILES CC1=C(C(=NN1)C)I

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 5 g
Formula Weight 222.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodo-3,5-dimethylpyrazole
RTECSNumber UQ6517000
TSCA No