CAS RN 2033-45-6
IUPAC Name:
4-iodo-3,5-dimethyl-1H-pyrazole
Synonyms:
4-iodo-3,5-dimethyl-1H-pyrazole
Molecular Weight (g/mol):
222.03
Molecular Formula:
C5H7IN2
InChi Key:
MZZXIXHKDJNBJQ-UHFFFAOYSA-N
SMILES:
CC1=C(I)C(C)=NN1
1
–
1
of
1
results
1
4-Iodo-3,5-dimethylpyrazole 98.0+%, TCI America™
CAS: 2033-45-6 Molecular Formula: C5H7IN2 Molecular Weight (g/mol): 222.029 MDL Number: MFCD00040247 InChI Key: MZZXIXHKDJNBJQ-UHFFFAOYSA-N Synonym: 3,5-dimethyl-4-iodopyrazole,4-iodo-3,5-dimethylpyrazole,3,5-dimethyl-4-iodo-1h-pyrazole,pyrazole, 3,5-dimethyl-4-iodo,3,5-dimethyl-4-lodopyrazole,1h-pyrazole, 4-iodo-3,5-dimethyl,zlchem 469,acmc-209f8u,5-23-05-00159 beilstein handbook reference,ksc490o8h PubChem CID: 97115 IUPAC Name: 4-iodo-3,5-dimethyl-1H-pyrazole SMILES: CC1=C(C(=NN1)C)I