missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodo-3,5-dimethylisoxazole 98.0+%, TCI America™
$57.79 - $199.46
Chemical Identifiers
| CAS | 10557-85-4 |
|---|---|
| Molecular Formula | C5H6INO |
| Molecular Weight (g/mol) | 223.01 |
| MDL Number | MFCD00173743 |
| InChI Key | NMNOXVWRJISEFE-UHFFFAOYSA-N |
| Synonym | 3,5-dimethyl-4-iodoisoxazole, 4-iodo-3,5-dimethylisoxazole, isoxazole, 4-iodo-3,5-dimethyl, 4-iodo-3,5-dimethyl-isoxazole, 3,5-dimethyl-4-iodoisorazole, pubchem3546, buttpark 97\06-60, acmc-2098gw, ksc496a2p, 3.5-dimethyl-4-iodoisorazole |
| PubChem CID | 613883 |
| IUPAC Name | 4-iodo-3,5-dimethyl-1,2-oxazole |
| SMILES | CC1=C(I)C(C)=NO1 |
Chemical Identifiers
| 10557-85-4 | |
| 223.01 | |
| NMNOXVWRJISEFE-UHFFFAOYSA-N | |
| 613883 | |
| CC1=C(I)C(C)=NO1 |
| C5H6INO | |
| MFCD00173743 | |
| 3,5-dimethyl-4-iodoisoxazole, 4-iodo-3,5-dimethylisoxazole, isoxazole, 4-iodo-3,5-dimethyl, 4-iodo-3,5-dimethyl-isoxazole, 3,5-dimethyl-4-iodoisorazole, pubchem3546, buttpark 97\06-60, acmc-2098gw, ksc496a2p, 3.5-dimethyl-4-iodoisorazole | |
| 4-iodo-3,5-dimethyl-1,2-oxazole |
Specifications
| 10557-85-4 | |
| C5H6INO | |
| 1 g | |
| NMNOXVWRJISEFE-UHFFFAOYSA-N | |
| 4-iodo-3,5-dimethyl-1,2-oxazole | |
| 613883 | |
| ≥98.0% (GC) | |
| 4-Iodo-3,5-dimethylisoxazole |
| 54°C | |
| MFCD00173743 | |
| 3,5-dimethyl-4-iodoisoxazole, 4-iodo-3,5-dimethylisoxazole, isoxazole, 4-iodo-3,5-dimethyl, 4-iodo-3,5-dimethyl-isoxazole, 3,5-dimethyl-4-iodoisorazole, pubchem3546, buttpark 97\06-60, acmc-2098gw, ksc496a2p, 3.5-dimethyl-4-iodoisorazole | |
| CC1=C(I)C(C)=NO1 | |
| 223.01 | |
| 223.01 | |
| Crystalline Powder |
Safety and Handling
TSCA : No