CAS RN 10557-85-4
IUPAC Name:
4-iodo-3,5-dimethyl-1,2-oxazole
Synonyms:
4-iodo-3,5-dimethyl-1,2-oxazole
Molecular Weight (g/mol):
223.01
Molecular Formula:
C5H6INO
InChi Key:
NMNOXVWRJISEFE-UHFFFAOYSA-N
SMILES:
CC1=C(I)C(C)=NO1
1
–
1
of
1
results
1
4-Iodo-3,5-dimethylisoxazole 98.0+%, TCI America™
CAS: 10557-85-4 Molecular Formula: C5H6INO Molecular Weight (g/mol): 223.01 MDL Number: MFCD00173743 InChI Key: NMNOXVWRJISEFE-UHFFFAOYSA-N Synonym: 3,5-dimethyl-4-iodoisoxazole,4-iodo-3,5-dimethylisoxazole,isoxazole, 4-iodo-3,5-dimethyl,4-iodo-3,5-dimethyl-isoxazole,3,5-dimethyl-4-iodoisorazole,pubchem3546,buttpark 97\06-60,acmc-2098gw,ksc496a2p,3.5-dimethyl-4-iodoisorazole PubChem CID: 613883 IUPAC Name: 4-iodo-3,5-dimethyl-1,2-oxazole SMILES: CC1=C(I)C(C)=NO1