missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-3,5-dimethylisoxazole 98.0+%, TCI America™

Catalog No. I07271G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07271G 1 g
I07275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07271G Supplier TCI America Supplier No. I07271G

Chemical Identifiers

CAS 10557-85-4
Molecular Formula C5H6INO
Molecular Weight (g/mol) 223.01
MDL Number MFCD00173743
InChI Key NMNOXVWRJISEFE-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-iodoisoxazole, 4-iodo-3,5-dimethylisoxazole, isoxazole, 4-iodo-3,5-dimethyl, 4-iodo-3,5-dimethyl-isoxazole, 3,5-dimethyl-4-iodoisorazole, pubchem3546, buttpark 97\06-60, acmc-2098gw, ksc496a2p, 3.5-dimethyl-4-iodoisorazole
PubChem CID 613883
IUPAC Name 4-iodo-3,5-dimethyl-1,2-oxazole
SMILES CC1=C(I)C(C)=NO1

Specifications

Melting Point 54°C
Quantity 1 g
Formula Weight 223.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodo-3,5-dimethylisoxazole