missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-2-methylaniline 98.0+%, TCI America™

Catalog No. I06905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06905G 5 g
I069025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06905G Supplier TCI America Supplier No. I06905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13194-68-8
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.052
MDL Number MFCD00025299
InChI Key BGKLFAQCHHCZRZ-UHFFFAOYSA-N
Synonym 2-amino-5-iodotoluene, 4-iodo-2-methylbenzenamine, 4-iodo-o-toluidine, benzenamine, 4-iodo-2-methyl, 4-iodo-2-methyl-phenylamine, 2-methyl-4-iodoaniline, 4-iodo-2-methylphenylamine, 4-iodo-2-methyl-aniline, aniline, 4-iodo-2-methyl, aurora 1126
PubChem CID 83221
IUPAC Name 4-iodo-2-methylaniline
SMILES CC1=C(C=CC(=C1)I)N

Specifications

Melting Point 88°C
Color Purple
Quantity 5 g
Formula Weight 233.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodo-2-methylaniline