missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-2,5-dimethylphenol, 98%

Catalog No. AAH5056503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5056503 1 g
AAH5056506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5056503 Supplier Thermo Scientific Chemicals Supplier No. H5056503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 114971-53-8 | C8H9IO | 248.063 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 114971-53-8
Molecular Formula C8H9IO
Molecular Weight (g/mol) 248.063
MDL Number MFCD00219922
InChI Key RFAXNKKOQUYDKE-UHFFFAOYSA-N
Synonym 2,5-dimethyl-4-iodophenol, 4-iodo-2,5-dimethyl-phenol, phenol,4-iodo-2,5-dimethyl, acmc-20aoag, 2-hydroxy-5-iodo-p-xylene, 4-iodanyl-2,5-dimethyl-phenol
PubChem CID 2736261
IUPAC Name 4-iodo-2,5-dimethylphenol
SMILES CC1=CC(=C(C=C1I)C)O

Specifications

Quantity 1 g
Sensitivity Light sensitive
Formula Weight 248.06
Percent Purity 98%
Chemical Name or Material 4-Iodo-2,5-dimethylphenol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only