missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-1-trityl-1H-imidazole, 98%, Thermo Scientific Chemicals

Catalog No. AC459200010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459200010 1 g
AC459200050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459200010 Supplier Thermo Scientific Chemicals Supplier No. 459200010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96797-15-8
Molecular Formula C22H17IN2
Molecular Weight (g/mol) 436.30
MDL Number MFCD02179542
InChI Key DXJZJYPLPZEYBH-UHFFFAOYSA-N
Synonym 4-iodo-1-trityl-1h-imidazole, 4-iodo-1-triphenylmethyl imidazole, 1-trityl-4-iodoimidazole, 4-iodo-1-triphenylmethyl-1h-imidazole, 4-iodo-1-trityl-imidazole, 1h-imidazole, 4-iodo-1-triphenylmethyl, pubchem8995, 4-iodo-l-tritylimidazole, n1-trityl-4-iodoimidazole
PubChem CID 618252
IUPAC Name 4-iodo-1-tritylimidazole
SMILES IC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 223.0°C to 227.5°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 436.3
Percent Purity 98%
Chemical Name or Material 4-Iodo-1-trityl-1H-imidazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only