missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-1-(triphenylmethyl)imidazole 98.0+%, TCI America™

Catalog No. I08901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08901G 1 g
I08905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08901G Supplier TCI America Supplier No. I08901G

Chemical Identifiers

CAS 96797-15-8
Molecular Formula C22H17IN2
Molecular Weight (g/mol) 436.30
MDL Number MFCD02179542
InChI Key DXJZJYPLPZEYBH-UHFFFAOYSA-N
Synonym 4-iodo-1-trityl-1h-imidazole, 4-iodo-1-triphenylmethyl imidazole, 1-trityl-4-iodoimidazole, 4-iodo-1-triphenylmethyl-1h-imidazole, 4-iodo-1-trityl-imidazole, 1h-imidazole, 4-iodo-1-triphenylmethyl, pubchem8995, 4-iodo-l-tritylimidazole, n1-trityl-4-iodoimidazole
PubChem CID 618252
IUPAC Name 4-iodo-1-(triphenylmethyl)-1H-imidazole
SMILES IC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 224°C
Color White
Quantity 1 g
Formula Weight 436.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodo-1-(triphenylmethyl)imidazole