missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-1-phenyl-1H-pyrazole, 95%

Catalog No. AAH32612MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32612MD 250 mg
AAH3261203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32612MD Supplier Thermo Scientific Chemicals Supplier No. H32612MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23889-85-2 | C9H7IN2 | 270.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23889-85-2
Molecular Formula C9H7IN2
Molecular Weight (g/mol) 270.073
MDL Number MFCD11707168
InChI Key PYJVDCYRDQUUKO-UHFFFAOYSA-N
Synonym 4-iodo-1-phenyl-1h-pyrazole, 1-phenyl-4-iodopyrazole, 1h-pyrazole, 4-iodo-1-phenyl, pubchem13298, 4-iodanyl-1-phenyl-pyrazole, 1-phenyl-4-iodo-1h-pyrazole
PubChem CID 14972076
IUPAC Name 4-iodo-1-phenylpyrazole
SMILES C1=CC=C(C=C1)N2C=C(C=N2)I

Specifications

Quantity 250 mg
Formula Weight 270.07
Percent Purity 95%
Chemical Name or Material 4-Iodo-1-phenyl-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only