CAS RN 23889-85-2
IUPAC Name:
4-iodo-1-phenyl-1H-pyrazole
Synonyms:
4-iodo-1-phenylpyrazole
Molecular Weight (g/mol):
270.07
Molecular Formula:
C9H7IN2
InChi Key:
PYJVDCYRDQUUKO-UHFFFAOYSA-N
SMILES:
IC1=CN(N=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
4-Iodo-1-phenyl-1H-pyrazole, 95%
CAS: 23889-85-2 Molecular Formula: C9H7IN2 Molecular Weight (g/mol): 270.073 MDL Number: MFCD11707168 InChI Key: PYJVDCYRDQUUKO-UHFFFAOYSA-N Synonym: 4-iodo-1-phenyl-1h-pyrazole,1-phenyl-4-iodopyrazole,1h-pyrazole, 4-iodo-1-phenyl,pubchem13298,4-iodanyl-1-phenyl-pyrazole,1-phenyl-4-iodo-1h-pyrazole PubChem CID: 14972076 IUPAC Name: 4-iodo-1-phenylpyrazole SMILES: C1=CC=C(C=C1)N2C=C(C=N2)I