missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxytamoxifen, (E) and (Z) isomers (50:50), MilliporeSigma™ Supelco™

Catalog No. 111015166
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015166 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015166 Supplier Merck Emd Millipore Supplier No. T17610MG
Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 68392-35-8
Physical Form Powder
Linear Formula C26H29 NO2
Synonym 4-(1-[4-(Dimethylaminoethoxy)phenyl]-2-phenyl-1-butenyl)phenol; 4-OHT; cis/trans-4-Hydroxytamoxifen
Solubility Soluble in ethanol
Type Analytical Standard
Quantity 10 mg
Formula Weight 387.51
Grade Analytical Standard