missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxytamoxifen (E) and (Z) isomers (50:50), ≥97.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015165
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015165 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015165 Supplier Merck Emd Millipore Supplier No. 9487310MG

4-Hydroxytamoxifen is a first generation, selective estrogen receptor modulator (SERM) that functions as an antagonist in breast cancer cells but can display estrogen-like activities in the uterus and bone.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 68392-35-8
Physical Form Neat
Linear Formula C26H29 NO2
Synonym 4-(1-[4-(Dimethylaminoethoxy)phenyl]-2-phenyl-1-butenyl)phenol; 4-OHT; cis/trans-4-Hydroxytamoxifen
Type Analytical Standard
Quantity 10 mg
Formula Weight 387.51
Percent Purity ≥97.5% (HPLC)
Grade Analytical Standard