missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxytamoxifen, Analytical Standard, (E) and (Z) isomers (50:50), MilliporeSigma™ Supelco™

Catalog No. 111015167
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015167 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015167 Supplier Merck Emd Millipore Supplier No. T17650MG

Specifications

Packaging Poly Bottle
CAS 68392-35-8
Physical Form Powder
Linear Formula C26H29NO2
Synonym 4-(1-[4-(Dimethylaminoethoxy)phenyl]-2-phenyl-1-butenyl)phenol; 4-OHT; cis/trans-4-Hydroxytamoxifen
Solubility Soluble in ethanol
Quantity 50 mg
Formula Weight 387.51
Grade Analytical Standard