missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxypyridine-3-sulfonic Acid 98.0+%, TCI America™
Supplier: TCI America H096325G
Specifications
| 4-Hydroxypyridine-3-sulfonic Acid | |
| C5H5NO4S | |
| 25 g | |
| AZRCSTMIVMQIPR-UHFFFAOYSA-N | |
| 4-oxo-1H-pyridine-3-sulfonic acid | |
| 3016673 | |
| ≥98.0% (HPLC,T) |
| 51498-37-4 | |
| MFCD02093473 | |
| 4-hydroxypyridine-3-sulfonic acid, 4-hydroxy-3-pyridinesulfonic acid, 4-hydroxypyridine-3-sulphonic acid, 4-hydroxy-3-pyridinesulphonic acid, 4-hydroxypyridie-3-sulphonic acid, hydroxypyridinesulfonicacid, 3-pyridinesulfonic acid, 4-hydroxy, acmc-209ku1, 4-hydroxy-3-pyridinesulfonic, 4-pyridinol-3-sulfonic acid | |
| C1=CNC=C(C1=O)S(=O)(=O)O | |
| 175.158 | |
| 175.16 | |
| Crystalline Powder |
Chemical Identifiers
| 51498-37-4 | |
| 175.158 | |
| AZRCSTMIVMQIPR-UHFFFAOYSA-N | |
| 3016673 | |
| C1=CNC=C(C1=O)S(=O)(=O)O |
| C5H5NO4S | |
| MFCD02093473 | |
| 4-hydroxypyridine-3-sulfonic acid, 4-hydroxy-3-pyridinesulfonic acid, 4-hydroxypyridine-3-sulphonic acid, 4-hydroxy-3-pyridinesulphonic acid, 4-hydroxypyridie-3-sulphonic acid, hydroxypyridinesulfonicacid, 3-pyridinesulfonic acid, 4-hydroxy, acmc-209ku1, 4-hydroxy-3-pyridinesulfonic, 4-pyridinol-3-sulfonic acid | |
| 4-oxo-1H-pyridine-3-sulfonic acid |
Safety and Handling
TSCA : No