accessibility menu, dialog, popup

UserName

CAS RN 51498-37-4

IUPAC Name: 4-oxo-1,4-dihydropyridine-3-sulfonic acid
Synonyms: 4-oxo-1H-pyridine-3-sulfonic acid
Molecular Weight (g/mol): 175.16
Molecular Formula: C5H5NO4S
InChi Key: AZRCSTMIVMQIPR-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=CNC=CC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results