missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxypyridine-3-sulfonic Acid 98.0+%, TCI America™

Catalog No. H09635G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09635G 5 g
H096325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09635G Supplier TCI America Supplier No. H09635G

Chemical Identifiers

CAS 51498-37-4
Molecular Formula C5H5NO4S
Molecular Weight (g/mol) 175.158
MDL Number MFCD02093473
InChI Key AZRCSTMIVMQIPR-UHFFFAOYSA-N
Synonym 4-hydroxypyridine-3-sulfonic acid, 4-hydroxy-3-pyridinesulfonic acid, 4-hydroxypyridine-3-sulphonic acid, 4-hydroxy-3-pyridinesulphonic acid, 4-hydroxypyridie-3-sulphonic acid, hydroxypyridinesulfonicacid, 3-pyridinesulfonic acid, 4-hydroxy, acmc-209ku1, 4-hydroxy-3-pyridinesulfonic, 4-pyridinol-3-sulfonic acid
PubChem CID 3016673
IUPAC Name 4-oxo-1H-pyridine-3-sulfonic acid
SMILES C1=CNC=C(C1=O)S(=O)(=O)O

Specifications

Quantity 5 g
Formula Weight 175.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxypyridine-3-sulfonic Acid