missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxypyridine-2-carboxylic acid, 97%

Catalog No. p-7032698
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34262MD 250 mg
AAH3426203 1 g
AAH3426206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH34262MD Supplier Thermo Scientific Chemicals Supplier No. H34262MD
Only null left

CAS: 22468-26-4 | C6H5NO3 | 139.11 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
slightly soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 22468-26-4
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00521155,MFCD11656334
InChI Key MXXLHBCSVDDTIX-UHFFFAOYSA-N
Synonym 4-hydroxypicolinic acid, 4-hydroxypyridine-2-carboxylic acid, 4-hydroxy-2-pyridinecarboxylic acid, 4-oxo-1,4-dihydropyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 4-hydroxy, 4-hydroxypyridine-2-carboxylicacid, 2-pyridinecarboxylic acid, 1,4-dihydro-4-oxo, 4-hydroxy-picolinsaure, pubchem17131, acmc-209fwn
PubChem CID 735149
IUPAC Name 4-oxo-1H-pyridine-2-carboxylic acid
SMILES OC(=O)C1=CC(=O)C=CN1

Specifications

Melting Point ∽280°C (decomposition)
Quantity 250 mg
Solubility Information slightly soluble in water.
Formula Weight 139.11
Percent Purity 97%
Chemical Name or Material 4-Hydroxypyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only