accessibility menu, dialog, popup

UserName

NHOOHO

CAS RN 22468-26-4

IUPAC Name: 4-oxo-1,4-dihydropyridine-2-carboxylic acid
Synonyms: 4-oxo-1H-pyridine-2-carboxylic acid
Molecular Weight (g/mol): 139.11
Molecular Formula: C6H5NO3
InChi Key: MXXLHBCSVDDTIX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=O)C=CN1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)