missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxypropiophenone 98.0+%, TCI America™

Catalog No. p-7130473
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
H029925G 25 g
H0299500G 500 g
2 options

Catalog No. H029925G

Supplier: TCI America H029925G

Only null left

Chemical Identifiers

CAS 70-70-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002361
InChI Key RARSHUDCJQSEFJ-UHFFFAOYSA-N
Synonym 4'-hydroxypropiophenone, paroxypropione, 4-hydroxypropiophenone, 1-4-hydroxyphenyl propan-1-one, mepal, paroxypropion, frenantol, frenohypon, p-hydroxypropiophenone, proxiphenon
PubChem CID 6271
ChEBI CHEBI:32352
IUPAC Name 1-(4-hydroxyphenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 150°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hydroxypropiophenone
EINECSNumber (9)-2077
RTECSNumber UH1925000
TSCA Yes