missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphthalic acid, 98%

Catalog No. p-7022360
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1509106 5 g
AAA1509114 25 g
AAA1509122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1509106 Supplier Thermo Scientific Chemicals Supplier No. A1509106
Only null left

CAS: 610-35-5 | C8H6O5 | 182.131 g/mol

4-Hydroxyphthalic acid is used as a precursor to prepare 3,5-dichloro-4-hydroxyphthalic acid using the reagent aqueous acetic acid and chlorine. Further, it is also used as a starting material in the manufacture of N-(4-carboxyphenyl)-4-acetoxyphthalimide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxyphthalic acid is used as a precursor to prepare 3,5-dichloro-4-hydroxyphthalic acid using the reagent aqueous acetic acid and chlorine. Further, it is also used as a starting material in the manufacture of N-(4-carboxyphenyl)-4-acetoxyphthalimide.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 610-35-5
Molecular Formula C8H6O5
Molecular Weight (g/mol) 182.131
MDL Number MFCD00013984
InChI Key MWRVRCAFWBBXTL-UHFFFAOYSA-N
Synonym 1,2-benzenedicarboxylic acid, 4-hydroxy, 4-hydroxybenzene-1,2-dicarboxylic acid, 4-hydroxy-1,2-benzenedicarboxylic acid, 3,4-dicarboxyphenol, 4-hydroxy-phthalic acid, acmc-1avol, ksc357m7h, 4-hydroxyphthalic acid, 4-hydroxy-1,2-benzenedicarboxylicacid, 1,2-benzenedicarboxylicacid, 4-hydroxy
PubChem CID 11881
ChEBI CHEBI:27600
IUPAC Name 4-hydroxyphthalic acid
SMILES C1=CC(=C(C=C1O)C(=O)O)C(=O)O

Specifications

Melting Point ∼190°C (decomposition)
Quantity 5 g
Solubility Information Slightly miscible with water.
Formula Weight 182.13
Percent Purity 98%
Chemical Name or Material 4-Hydroxyphthalic acid
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 210-221-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only