missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxyphenylpyruvic Acid 95.0+%, TCI America™
Specifications
Specifications
| Quantity | 5 g |
| Formula Weight | 180.16 |
| Percent Purity | ≥95.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Hydroxyphenylpyruvic Acid |
Safety and Handling
| RTECSNumber | UZ1000000 |
| TSCA | No |
| Recommended Storage | Refrigerator |