missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylpyruvic Acid 95.0+%, TCI America™

Catalog No. H02941G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H02941G 1 g
H02945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H02941G Supplier TCI America Supplier No. H02941G

Chemical Identifiers

CAS 156-39-8
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00002591
InChI Key KKADPXVIOXHVKN-UHFFFAOYSA-N
Synonym 4-hydroxyphenylpyruvic acid, testacid, 3-4-hydroxyphenyl-2-oxopropanoic acid, p-hydroxyphenylpyruvic acid, testacide, 4-hydroxyphenylpyruvate, 3-4-hydroxyphenyl pyruvic acid, p-hydroxyphenyl pyruvic acid, 3-p-hydroxyphenyl pyruvic acid, 3-p-hydroxyphenyl-2-oxopropanoic acid
PubChem CID 979
ChEBI CHEBI:15999
IUPAC Name 3-(4-hydroxyphenyl)-2-oxopropanoic acid
SMILES C1=CC(=CC=C1CC(=O)C(=O)O)O

Specifications

Quantity 1 g
Formula Weight 180.16
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxyphenylpyruvic Acid
RTECSNumber UZ1000000
TSCA No
Recommended Storage Refrigerator