missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylacetonitrile, 97%

Catalog No. AAA1097103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1097103 1 g
AAA1097106 5 g
AAA1097114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1097103 Supplier Thermo Scientific Chemicals Supplier No. A1097103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14191-95-8 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14191-95-8
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00002383
InChI Key AYKYOOPFBCOXSL-UHFFFAOYSA-N
Synonym 4-hydroxybenzyl cyanide, 4-hydroxyphenylacetonitrile, 2-4-hydroxyphenyl acetonitrile, p-hydroxybenzyl cyanide, 4-hydroxyphenyl acetonitrile, p-hydroxyphenylacetonitrile, 4-hydroxybenzylcyanide, 4-hydroxybenzeneacetonitrile, benzeneacetonitrile, 4-hydroxy, acetonitrile, p-hydroxyphenyl
PubChem CID 26548
ChEBI CHEBI:16667
IUPAC Name 2-(4-hydroxyphenyl)acetonitrile
SMILES OC1=CC=C(CC#N)C=C1

Specifications

Melting Point 66°C to 71°C
Boiling Point 329°C to 331°C
Quantity 1 g
Beilstein 1934470
Formula Weight 133.15
Percent Purity 97%
Chemical Name or Material 4-Hydroxyphenylacetonitrile
Hazard Category H302+H312+H332-H315-H319-H335-H500
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 238-046-0
RTECSNumber AM0530000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only