missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylacetamide 98.0+%, TCI America™

Catalog No. p-7130763
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H085325G 25 g
H0853500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H085325G Supplier TCI America Supplier No. H085325G
Only null left

Chemical Identifiers

CAS 17194-82-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00017145
InChI Key YBPAYPRLUDCSEY-UHFFFAOYSA-N
Synonym 4-hydroxyphenylacetamide, 2-4-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, 4-hydroxybenzeneacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxyphenyl acetamide, unii-0hy0n4itn4
PubChem CID 86986
IUPAC Name 2-(4-hydroxyphenyl)acetamide
SMILES C1=CC(=CC=C1CC(=O)N)O

Specifications

Melting Point 177°C
Quantity 25 g
Formula Weight 151.17
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxyphenylacetamide
EINECSNumber (3)-3726
RTECSNumber AC3851500
TSCA Yes