missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenyl Methyl Sulfone 98.0+%, TCI America™

Catalog No. H13675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13675G 5 g
H136725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13675G Supplier TCI America Supplier No. H13675G
Only null left

Chemical Identifiers

CAS 14763-60-1
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00047806
InChI Key KECCFSZFXLAGJS-UHFFFAOYSA-N
Synonym 4-methylsulfonyl phenol, 4-methanesulfonylphenol, 4-methanesulfonyl-phenol, 4-hydroxyphenyl methyl sulfone, 4-methylsulphonylphenol, phenol, p-methylsulfonyl, phenol, 4-methylsulfonyl, p-hydroxyphenyl methyl sulfone, p-methylsulfonyl phenol, unii-f7bif7zqj9
PubChem CID 123350
IUPAC Name 4-methanesulfonylphenol
SMILES CS(=O)(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 95°C
Color White
Quantity 5 g
Formula Weight 172.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxyphenyl Methyl Sulfone
RTECSNumber SM1530000
TSCA No