missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenyl benzoate, 98%

Catalog No. AAL0935006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0935006 5 g
AAL0935014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0935006 Supplier Thermo Scientific Chemicals Supplier No. L0935006

CAS: 2444-19-1 | C13H10O3 | 214.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2444-19-1
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00053304
InChI Key JFAXJRJMFOACBO-UHFFFAOYSA-N
Synonym hydroquinone monobenzoate, p-hydroxyphenyl benzoate, unii-zcy7o225cp, 4-benzoyloxyphenol, 1,4-diphenol, 1-benzoate, benzoic acid 4-hydroxyphenyl ester, 4-hydroxyphenyl benzoate, zcy7o225cp, 4-hydroxyphenylbenzoate, 4-hydroxyphenylbenzoat
PubChem CID 75549
IUPAC Name (4-hydroxyphenyl) benzoate
SMILES OC1=CC=C(OC(=O)C2=CC=CC=C2)C=C1

Specifications

Melting Point 164°C to 167°C
Quantity 5 g
Beilstein 2050596
Formula Weight 214.22
Percent Purity 98%
Chemical Name or Material 4-Hydroxyphenyl benzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-479-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only