accessibility menu, dialog, popup

UserName

CAS RN 2444-19-1

IUPAC Name: 4-hydroxyphenyl benzoate
Synonyms: 4-hydroxyphenyl benzoate
Molecular Weight (g/mol): 214.22
Molecular Formula: C13H10O3
InChi Key: JFAXJRJMFOACBO-UHFFFAOYSA-N
SMILES: OC1=CC=C(OC(=O)C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results