missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenethyl alcohol, 98%

Catalog No. p-146641
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC182500050 5 g Glass bottle
AC182500250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC182500050 Supplier Thermo Scientific Chemicals Supplier No. 182500050
Only null left

CAS: 501-94-0 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 501-94-0
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00002902
InChI Key YCCILVSKPBXVIP-UHFFFAOYSA-N
Synonym tyrosol, 2-4-hydroxyphenyl ethanol, 4-hydroxyphenethyl alcohol, 4-2-hydroxyethyl phenol, 4-hydroxyphenylethanol, 4-hydroxybenzeneethanol, p-hydroxyphenethyl alcohol, benzeneethanol, 4-hydroxy, p-tyrosol, p-hydroxyphenylethanol
PubChem CID 10393
ChEBI CHEBI:1879
IUPAC Name 4-(2-hydroxyethyl)phenol
SMILES C1=CC(=CC=C1CCO)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 89.0°C to 92.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HOC6H4CH2CH2OH
Quantity 5 g
Beilstein 06, IV, 5936
Merck Index 15, 10022
Solubility Information Solubility in water: 124g/L (25°C). Other solubilities: soluble in methanol,ethanol,acetone,chloroform,,diethyl ether,petroleum ether
Formula Weight 138.17
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Hydroxyphenethyl alcohol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously wit
EINECSNumber 207-930-8
TSCA TSCA

RUO – Research Use Only